Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 2-ethyl-6-methylpyrazine
TRC 2-ethyl-6-methylpyrazine
TRC 2-ethyl-6-methylpyrazine
77978-663
:
LGC Standards
New Product
:
TRC 2-ethyl-6-methylpyrazine
77978-663
:
LGC Standards
:
TRC-B438208-10MG
New Product
:
CAS No.:
13925-03-6
Description:
2-ethyl-6-methylpyrazine
Size:
10 mg