Order Entry
Puerto Rico
ContactUsLinkComponent
Silica Comparator Round 0 to 1 ppm
Silica Comparator Round 0 to 1 ppm
Catalog # 77674-682
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Silica Comparator Round 0 to 1 ppm
Catalog # 77674-682
Supplier:  CHEMetrics
Supplier Number:  C-9001
New Product
CAS Number:  

Specifications

  • Chemical method:
    Heteropoly Blue

Specifications

About this item

C-9001 Low Range Silica Comparator is a component of K-9010 Silica CHEMets Visual Test Kit and is designed to be compared against the color formed in R-9010 Silica CHEMets Refill. The comparator's range is applicable to many industrial applications. Color standards are blue in color and cover a range of 0 to 1.0 ppm SiO₂