ContactUsLinkComponent
Monochloramine Accessory Solution Pack
Monochloramine Accessory Solution Pack
Catalog # 77674-568
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Monochloramine Accessory Solution Pack
Catalog # 77674-568
Supplier:  CHEMetrics
Supplier Number:  A-6802
New Product
CAS Number:  
Specifications
  • Chemical method:
    Direct Nesslerization
Specifications
About this item
The A-6802 accessory pack supplies enough of each solution to perform approximately 120 tests. Quantity per pack: Three 10 ml bottles of A-6804 Stabilizer Solution and three 10 ml bottles of A-6805 Catalyzer Solution.