Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 2-[(4-Chlorophenyl)methoxy]-N,N-dimethyl-ethanamine Hydrochloride
TRC 2-[(4-Chlorophenyl)methoxy]-N,N-dimethyl-ethanamine Hydrochloride
TRC 2-[(4-Chlorophenyl)methoxy]-N,N-dimethyl-ethanamine Hydrochloride
77979-885
:
LGC Standards
New Product
:
TRC 2-[(4-Chlorophenyl)methoxy]-N,N-dimethyl-ethanamine Hydrochloride
77979-885
:
LGC Standards
:
TRC-C364697-50MG
New Product
:
CAS No.:
1135085-85-6
Description:
2-[(4-Chlorophenyl)methoxy]-N,N-dimethyl-ethanamine Hydrochloride
Size:
50 mg