Lipomed Meperidine.HCl (Pethidine.HCl), 1 mg/ml in Methanol (as free base)
Catalog # 77976-204
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- CAS No.:50-13-5
- Description:Meperidine.HCl (Pethidine.HCl), 1 mg/ml in Methanol (as free base)
- Size:1 ml