Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC Dibucaine Hydrochloride
TRC Dibucaine Hydrochloride
77982-298
:
LGC Standards
New Product
:
TRC Dibucaine Hydrochloride
77982-298
:
LGC Standards
:
TRC-D426500-10MG
New Product
:
CAS No.:
61-12-1
Description:
Dibucaine Hydrochloride
Size:
10 mg