Paragon Scientific RTM15 Mineral Oil Rotational Viscosity
Catalog # 77975-967
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Description:RTM15, Mineral oil rotational viscosity
- Size:500 ml
- Shipping Conditions:RT
- Storage Temperature:RT
- Certification:ISO 17034
Specifications
About this item
- Standard type: Single component