Order Entry
Puerto Rico
ContactUsLinkComponent
Chloride Instrumental Test 0 to 40 ppm
Chloride Instrumental Test 0 to 40 ppm
Catalog # 77674-750
Supplier:  CHEMetrics
New Product
CAS Number:  
undefined
Chloride Instrumental Test 0 to 40 ppm
Catalog # 77674-750
Supplier:  CHEMetrics
Supplier Number:  K-2103
New Product
CAS Number:  

Specifications

  • Chemical method:
    Ferric thiocyanate
  • Application:
    Drinking water;Surface and saline water;Domestic and industrial waste water

Specifications

About this item

Vacu-vials Kits require the use of the V-2000 Photometer or a spectrophotometer capable of accepting a 13 mm diameter round cell. Instrument sold separately. Although the test kit contains 30 ampoules, a fresh reagent ampoule blank must be prepared for each series of tests; therefore, the number of samples that can be tested with each kit will vary from a maximum of 29 to a minimum of 15.

  • Range: 0 to 40.0 ppm Method: Ferric Thiocyanate
Delivery: Kit K-2103 comes in a cardboard box and contains everything needed to perform up to 29 tests (except distilled water): thirty ampoules, activator solution, 25 ml sample cup, ampoule blank, 1.0 ml syringe, instructions, and MSDS.