Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC p-Salicylic Acid
TRC p-Salicylic Acid
TRC p-Salicylic Acid
Supplier:
LGC Standards
New Product
Certificates
Expand image
TRC p-Salicylic Acid
Supplier:
LGC Standards
New Product
About this item
Specifications
CAS No.:
99-96-7
Specifications