$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC (E)-Fluvoxamine-d3 Maleate
TRC (E)-Fluvoxamine-d3 Maleate
77985-032
:
LGC Standards
New Product
:
TRC (E)-Fluvoxamine-d3 Maleate
77985-032
:
LGC Standards
:
TRC-F603502-1MG
New Product
:
CAS No.:
1185245-56-0
Description:
(E)-Fluvoxamine-d3 Maleate
Size:
1 mg