$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Azelastine Hydrochloride
TRC Azelastine Hydrochloride
:
LGC Standards
New Product
TRC Azelastine Hydrochloride
:
LGC Standards
New Product
CAS No.:
79307-93-0