$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC Calcifediol
TRC Calcifediol
:
LGC Standards
New Product
TRC Calcifediol
:
LGC Standards
New Product
CAS No.:
19356-17-3