TRC (11b,16a)-9-Fluoro-11,21,21-trihydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione
Supplier: LGC Standards
New Product
Certificates
TRC (11b,16a)-9-Fluoro-11,21,21-trihydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione
Supplier: LGC Standards
New Product
About this item
Specifications
- CAS No.:161740-69-8
Specifications
Frequently Bought Together
PEEK Tubing, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
Upchurch Scientific® PEEK™ Tubing, IDEX Health & Science
Supplier: Upchurch Scientific
Upchurch Scientific® Flat-Bottom Fittings, For Tubing Ø > ¹/₁₆" (1.6 mm) and ≤ ¹/₈" (3.2 mm), IDEX Health & Science
Supplier: Upchurch Scientific
Vaplock™ Chemical Exhaust Filters and Color Indicators, Cole-Parmer
Supplier: Antyila Scientific
Manual Crimpers and Decappers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Ultra Inert Liners for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Maintenance Kits for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Reciprocating Dissolution Screens, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Gas Tubing for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Solvent Bottles and Waste Cans for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Tubing for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Purged Packed Inlet for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
pH Gradient Buffers
Supplier: Thermo Scientific
CrossLab Ultra-Microcells for UV-Vis and UV-Vis-NIR Systems, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Full Flow Filters, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
FRITS REPLACEMENT TWIST 10UM/15UM PK20
Supplier: GLEN RESEARCH CORP MS
CrossLab Connector Tubes from Spray Chamber to Torch for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)