Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC 3-Dimethylamino-2-methylpropyl Chloride Hydrochloride
TRC 3-Dimethylamino-2-methylpropyl Chloride Hydrochloride
TRC 3-Dimethylamino-2-methylpropyl Chloride Hydrochloride
Catalog # 77983-072
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
TRC 3-Dimethylamino-2-methylpropyl Chloride Hydrochloride
Catalog # 77983-072
Supplier:
LGC Standards
Supplier Number:
TRC-D470795-50MG
New Product
CAS Number:
Specifications
CAS No.:
4261-67-0
Description:
3-Dimethylamino-2-methylpropyl Chloride Hydrochloride
Size:
50 mg
Specifications
About this item
Product Details
Product Details