TRC Phenyltin Trichloride
New Product
- CAS No.:1124-19-2
Frequently Bought Together
Manual Crimpers and Decappers, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Vaplock™ Chemical Exhaust Filters and Color Indicators, Cole-Parmer
: Antyila Scientific
CrossLab Vacuum Pump Supplies for GC/MS, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Dissolution Baskets, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Bond Elut™ Adapters and Caps, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Flame Ionization Detector (FID) for GC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Solvent Bottles and Waste Cans for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Injectors for Fully Demountable Torches, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Detectors for ICP-MS, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
FRITS REPLACEMENT TWIST 10UM/15UM PK20
: GLEN RESEARCH CORP MS
pH Gradient Buffers
: Thermo Scientific
CrossLab Tubing for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
InfinityLab Well Plates, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Tubing Kits for ICP-OES
: AGILENT TECHNOLOGIES, INC (CSD)
Disintegration Tubes, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Capillaries for HPLC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)