Paragon Scientific Base Number, 1 mg KOH/g Certified Value
Catalog # 77976-018
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Description:Base number, 1 mg KOH/g certified value
- Size:3×125 g
- Storage Temperature:RT
Specifications
About this item
- Shipping conditions: Room temperature