Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
TRC [(Z)-Prop-1-enyl]phosphonic Acid (Technical Grade)
TRC [(Z)-Prop-1-enyl]phosphonic Acid (Technical Grade)
TRC [(Z)-Prop-1-enyl]phosphonic Acid (Technical Grade)
77995-449
:
LGC Standards
New Product
:
TRC [(Z)-Prop-1-enyl]phosphonic Acid (Technical Grade)
77995-449
:
LGC Standards
:
TRC-Z192700-1G
New Product
:
CAS No.:
25383-06-6
Description:
[(Z)-Prop-1-enyl]phosphonic Acid (Technical Grade)
Size:
1 g