ContactUsLinkComponent
CHEMets® Chlorine Hypochlorite Visual Test Kit 0 to 1.55%
  76176-698
New Product
 :  
undefined
CHEMets® Chlorine Hypochlorite Visual Test Kit 0 to 1.55%
  76176-698
 :  K-5808
New Product
 :  
  • Chemical method:
    DPD (hypochlorite)
  • Application:
    Drinking water;Waste water;Surface and ground water
Range: 0 to 1.55% as NaOClMDL: 0.3%. Method: DPD (hypochlorite).
 : Kit comes in a plastic case and contains everything needed to perform 30 Chlorine (hypochlorite) tests (except distilled water):refill, comparator, 25 ml sample cup, sample Prep Cup, 3.0 ml syringe, 200 µl MiniPet® and instructions.