$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
TRC 3-Difluoromethyl-1-methylpyrazole-4-carboxylic Acid Ethyl Ester
TRC 3-Difluoromethyl-1-methylpyrazole-4-carboxylic Acid Ethyl Ester
77982-682
:
LGC Standards
New Product
:
TRC 3-Difluoromethyl-1-methylpyrazole-4-carboxylic Acid Ethyl Ester
77982-682
:
LGC Standards
:
TRC-D446860-10G
New Product
:
CAS No.:
141573-95-7
Description:
3-Difluoromethyl-1-methylpyrazole-4-carboxylic Acid Ethyl Ester
Size:
10 g