ContactUsLinkComponent
NeutraGreen Aldehyde Test Strips
NeutraGreen Aldehyde Test Strips
Catalog # 77908-090
Supplier:  StatLab
New Product
CAS Number:  
NeutraGreen Aldehyde Test Strips
Catalog # 77908-090
Supplier:  StatLab
Supplier Number:  NTS-50
New Product
CAS Number:  
Specifications
  • Description:
    NeutraGreen aldehyde test strips
Specifications
About this item
NeutraGreen test strips are used to help determine if waste formalin has been safely and effectively neutralized for drain disposal.
A quick dip providing color-coded results is an effective and simple way to test. Now available in a new, smaller pack size of 50.