CytomeGallonovirus (CMV) IHC Control Slides
Catalog # 77907-890
Supplier: StatLab
New Product
Specifications
- Description:CytomeGallonovirus (CMV) IHC control slides
Specifications
About this item
Cytomegalovirus (CMV) IHC control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
Stained slide is not included
Tissue: known positive
Special stain or antibody: CMV
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.