ContactUsLinkComponent
Small Intestine Control Slides
Small Intestine Control Slides
Catalog # 77907-968
Supplier:  StatLab
New Product
undefined
Small Intestine Control Slides
Catalog # 77907-968
Supplier:  StatLab
Supplier Number:  CSS0325P
New Product
Specifications
  • Description:
    Small Intestine control slides
Specifications
About this item
Small Intestine control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
  • Stained slide is not included
    Tissue: Small Intestine
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90° corners. Each package includes specific staining protocol.