Scored StatPrep Cervical Brush, Sterile
77907-862
: StatLab
New Product
- Description:Scored StatPrep cervical brush, sterile
Scored brush intended for use in liquid based cytology endocervical cell collection.
Sterile 8" length
Individually packed