PathFlow Supa Mega Slim Cassettes
About this item
The Supa Mega Slim cassette is being widely adopted in clinical, research, and toxicology laboratories.
- Casting can be done in 20 minutes at −8 °C
- Large color range available
- Specific base molds
The Supa Mega Slim cassette offers better reagent flow and improved turnaround times, with less wax used during the embedding process.
6 Options Available Below
Product Details & Documents
The Supa Mega Slim cassette is being widely adopted in clinical, research, and toxicology laboratories.
- Casting can be done in 20 minutes at −8 °C
- Large color range available
- Specific base molds
The Supa Mega Slim cassette offers better reagent flow and improved turnaround times, with less wax used during the embedding process.
Product Family Options
Product Information
- ColorAvailabilityPrice
- 77908-014PathFlow supa mega slim cassettesSupplier Number: EAN-0102-02ASpecifications:New ProductNew Product
- 77908-016PathFlow supa mega slim cassettesSupplier Number: EAN-0104-02ASpecifications:New ProductNew Product
- 77908-018PathFlow supa mega slim cassettesSupplier Number: EAN-0106-02ASpecifications:New ProductNew Product
- 77908-020PathFlow supa mega slim cassettesSupplier Number: EAN-0108-02ASpecifications:New ProductNew Product
- 77908-022PathFlow supa mega slim cassettesSupplier Number: EAN-0109-02ASpecifications:New ProductNew Product
- 77908-024PathFlow supa mega slim cassettesSupplier Number: EAN-0110-02ASpecifications:New ProductNew Product