About this item
Economical interlocking corrugated storage for slides and cassettes.
Tabs allow stacking
Available as a standard 4-drawer file, or larger capacity 8-drawer file
When used as a 4-drawer file, holds approximately 2000 slides. When used as an 8-drawer file holds approximately 500 tissue blocks.