Large Lab Wipe
Large lab wipes, 11×13"
Catalog # 77908-138
New Product
Supplier: StatLab
Supplier #: SL5790
Jump to:
Product Details & Documents
Use as an absorbent work surface for coverslipping or for general lab cleanup.
- Low-linting
- Heavy-duty, air-layered material
Specifications
- Description:Large lab wipes
- L×W:11×13"