Gram Stain +/- Control Slides
Catalog # 77907-912
Supplier: StatLab
New Product
Specifications
- Description:Gram stain ± control slides
Specifications
About this item
Gram Stain +/- control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
Includes 1 stained slide Tissue: known positive
Special stain or antibody: Gram stain
Use these slides with confidence to evaluate stain quality and performance in histology laboratories. StatLab control slides are created using Millennia adhesion white glass slides with 90 degree corners. Each pack includes specific staining protocol.