Disposable Scalpel, #22
77907-972
: StatLab
New Product
- Sterility:Sterile
- Description:Disposable scalpel, #22
One-piece design incorporates a stainless steel blade on a rigid handle.
Disposable #22 scalpel
Sterile 10/box
Each scalpel is supplied with a flexible blade cover for the end user’s protection.