Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
(1S,2R,3R)-Aprepitant, TLC Pharmaceutical Standards
(1S,2R,3R)-Aprepitant, TLC Pharmaceutical Standards
:
TLC Standards
New Product
(1S,2R,3R)-Aprepitant, TLC Pharmaceutical Standards
:
TLC Standards
New Product
CAS No.:
1242175-34-3