Anti-SBDS Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
New Product
Type:
Antigen: SBDS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77846-347
- 77846-348
- Antigen Symbol:
- SBDS
- SBDS
- Antigen Synonyms:
- 4733401P19Rik|AI836084|CGI 97|CGI-97|FLJ10917|MGC105922|Protein 22A3|Ribosome maturation protein SBDS_HUMAN|SDS|Shwachman Bodian Diamond syndrome protein|Shwachman Bodian Diamond syndrome protein homolog|Shwachman Bodian-Diamond syndrome|Shwachman-Bodian-Diamond syndrome protein|SWDS
- 4733401P19Rik|AI836084|CGI 97|CGI-97|FLJ10917|MGC105922|Protein 22A3|Ribosome maturation protein SBDS_HUMAN|SDS|Shwachman Bodian Diamond syndrome protein|Shwachman Bodian Diamond syndrome protein homolog|Shwachman Bodian-Diamond syndrome|Shwachman-Bodian-Diamond syndrome protein|SWDS
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y3A5
- Q9Y3A5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Ribosome maturation protein SBDS protein (37-153AA)
- Recombinant Human Ribosome maturation protein SBDS protein (37-153AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice