Anti-CHMP2B Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: CHMP2B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-721
- 77844-720
- Antigen Symbol:
- CHMP2B
- CHMP2B
- Antigen Synonyms:
- ALS17|Charged multivesicular body protein 2b|CHM2B_HUMAN|CHMP family|member 2B|CHMP2.5|CHMP2b|Chromatin modifying protein 2b|Chromatin-modifying protein 2b|DMT1|hVps2-2|Vacuolar protein sorting 2|yeast|homolog of|B|Vacuolar protein sorting 2-2|Vacuolar protein sorting-associated protein 2-2|VPS2 homolog B|Vps2-2|VPS2B
- ALS17|Charged multivesicular body protein 2b|CHM2B_HUMAN|CHMP family|member 2B|CHMP2.5|CHMP2b|Chromatin modifying protein 2b|Chromatin-modifying protein 2b|DMT1|hVps2-2|Vacuolar protein sorting 2|yeast|homolog of|B|Vacuolar protein sorting 2-2|Vacuolar protein sorting-associated protein 2-2|VPS2 homolog B|Vps2-2|VPS2B
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UQN3
- Q9UQN3
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Charged multivesicular body protein 2b protein (1-213AA)
- Recombinant Human Charged multivesicular body protein 2b protein (1-213AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice