Anti-P4HB Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77851-783
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:P4HB
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P07237
- Antigen Synonyms:Cellular thyroid hormone binding protein|Cellular thyroid hormone-binding protein|Collagen prolyl 4 hydroxylase beta|Disulphide Isomerase|DSI|EC 5.3.4.1|Endoplasmic reticulum resident protein 59|ER protein 59|ERBA2L|ERp59|GIT|Gltathione insulin transhydrogenase|Glutathione insulin transhydrogenase|P4Hbeta|p55|PDI|PDIA1|PDIA1_HUMAN|PDIR|PHDB|PO4DB|PO4HB|Procollagen proline 2 oxoglutarate 4 dioxygenase (proline 4 hydroxylase) beta polypeptide (protein disulfide isomerase associated 1)|Procollagen proline 2 oxoglutarate 4 dioxygenase beta subunit|PROHB|Prolyl 4 hydroxylase beta polypeptide|Prolyl 4 hydroxylase beta subunit|Prolyl 4 hydroxylase subunit beta|Prolyl 4-hydroxylase subunit beta|Protein disulfide isomerase associated 1|Protein disulfide isomerase, family A, member 1|Protein disulfide isomerase/oxidoreductase|Protein disulfide-isomerase|Protocollagen hydroxylase|Thbp|Thyroid hormone binding protein p55|Thyroid hormone binding protein p55 cellular|V erb a avian erythroblastic leukemia viral oncogene homolog 2 like
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Protein disulfide-isomerase protein (29-508AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human