Anti-UCHL3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:2000 to 1:5000, WB:1:500 to 1:2000, IHC:1:25 to 1:100
Type:
Antigen: UCHL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77850-087
- 77850-088
- Antigen Symbol:
- UCHL3
- UCHL3
- Antigen Synonyms:
- Ubiquitin carboxyl-terminal hydrolase isozyme L3|Ubiquitin thioesterase L3|Ubiquitin thiolesterase|Ubiquitin thiolesterase L3|UCH L3|UCH-L3|UCHL3_HUMAN
- Ubiquitin carboxyl-terminal hydrolase isozyme L3|Ubiquitin thioesterase L3|Ubiquitin thiolesterase|Ubiquitin thiolesterase L3|UCH L3|UCH-L3|UCHL3_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P15374
- P15374
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Full length fusion protein
- Full length fusion protein
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice