Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-ART1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Anti-ART1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77851-447
Supplier:  AFG Bioscience
New Product
undefined
Anti-ART1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77851-447
Supplier:  AFG Bioscience
Supplier Number:  A13222-50UL
New Product

Specifications

  • Antigen Symbol:
    ART1
  • Clonality:
    Recombinant Polyclonal
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Size:
    50 µl
  • Form:
    Liquid
  • Gene ID:
    P52961
  • Antigen Synonyms:
    ART1GPI-linked NAD(P)(+)--arginine ADP-ribosyltransferase 1|EC 2.4.2.31|ADP-ribosyltransferase C2 and C3 toxin-like 1|ARTC1|Mono(ADP-ribosyl)transferase 1|CD antigen CD296
  • Storage Buffer:
    0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage Temperature:
    Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
  • Shipping Temperature:
    Shipped on blue ice
  • Immunogen:
    Recombinant Human GPI-linked NAD(P)(+)--arginine ADP-ribosyltransferase 1 protein (46-142AA )
  • Purification:
    >95%, Protein G purified
  • Packaging:
    Vial

Specifications

About this item

Type:
Antigen: ART1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human