Anti-ERCC6L2 Rabbit Recombinant Polyclonal Antibody
77852-185
New Product
- Antigen Symbol:ERCC6L2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q5T890
- Antigen Synonyms:C9orf102|Chromosome 9 open reading frame 102|excision repair cross-complementing rodent repair deficiency|complementation group 6-like 2|FLJ37706|MGC30192|MGC43364|Putative DNA repair and recombination protein RAD26-like|putative repair and recombination helicase RAD26L|RAD26_HUMAN|Rad26l|RAD26L hypothetical protein|SR278|stretch responsive protein 278
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human DNA excision repair protein ERCC-6-like 2 protein (72-371AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: ERCC6L2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human