Anti-CCT8 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77852-602
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CCT8
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P50990
- Antigen Synonyms:C21orf112|CCTQ|KIAA0002|T-complex protein 1 subunit theta|TCP-1-theta|CCT-theta|Chaperonin containing T-complex polypeptide 1 subunit 8|Renal carcinoma antigen NY-REN-15
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human T-complex protein 1 subunit theta protein (224-491AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CCT8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human