Anti-ADA Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
77850-718
New Product
- Antigen Symbol:ADA
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P00813
- Antigen Synonyms:ADA_HUMAN|ADA1|Adenosine aminohydrolase|Adenosine deaminase
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Adenosine deaminase protein (157-343AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: ADA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human