Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-DCTN4 Rabbit Recombinant Polyclonal Antibody
  77845-073
New Product
undefined
Anti-DCTN4 Rabbit Recombinant Polyclonal Antibody
  77845-073
 :  A71807-50UG
New Product

 

  • Antigen Symbol:
    DCTN4
  • Clonality:
    Recombinant Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    50 µg
  • Form:
    Liquid
  • Gene ID:
    Q9UJW0
  • Antigen Synonyms:
    DCTN 4| Dctn4| DCTN4_HUMAN| Dynactin 4 (p62)| Dynactin 4| Dynactin p62 subunit| Dynactin subunit 4| Dynactin subunit p62| Dynactin4| p62
  • Storage Buffer:
    0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
  • Storage Temperature:
    Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
    Shipped on blue ice
  • Immunogen:
    Recombinant Human Dynactin subunit 4 protein (2-460AA)
  • Purification:
    >95%, Protein G purified
  • Packaging:
    Vial

 

 

Recommended dilution: WB:1:1000 to 1:5000

Type:
Antigen: DCTN4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human