Anti-TRAF3 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:1000 to 1:5000, WB:1:200 to 1:1000, IHC:1:25 to 1:100
Type:
Antigen: TRAF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77848-189
- 77848-188
- Antigen Symbol:
- TRAF3
- TRAF3
- Antigen Synonyms:
- CAP 1|CAP-1|CAP1|CD40 associated protein 1|CD40 binding protein|CD40 bp|CD40 receptor associated factor 1|CD40 receptor-associated factor 1|CD40-binding protein|CD40BP|CRAF 1|CRAF1|IIAE5|LAP 1|LAP1|LMP1 associated protein|LMP1-associated protein 1|MIPT3|TNF receptor associated factor 3|TNF receptor-associated factor 3|TRAF 3|Traf3|TRAF3_HUMAN
- CAP 1|CAP-1|CAP1|CD40 associated protein 1|CD40 binding protein|CD40 bp|CD40 receptor associated factor 1|CD40 receptor-associated factor 1|CD40-binding protein|CD40BP|CRAF 1|CRAF1|IIAE5|LAP 1|LAP1|LMP1 associated protein|LMP1-associated protein 1|MIPT3|TNF receptor associated factor 3|TNF receptor-associated factor 3|TRAF 3|Traf3|TRAF3_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q13114
- Q13114
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human TRAF3
- Synthetic peptide of Human TRAF3
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice