Anti-DNAJC6 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
77854-264
New Product
- Antigen Symbol:DNAJC6
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O75061
- Antigen Synonyms:KIAA0473|Putative tyrosine-protein phosphatase auxilin|EC 3.1.3.48|DnaJ homolog subfamily C member 6
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Putative tyrosine-protein phosphatase auxilin protein (201-500AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Type:
Antigen: DNAJC6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human