Anti-PCDHB16 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IF: 1:50 to 1:200
Type:
Antigen: PCDHB16
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-450
- 77843-449
- Antigen Symbol:
- PCDHB16
- PCDHB16
- Antigen Synonyms:
- Cadherin ME1|KIAA1621|ME 1|ME1|PCDBG_HUMAN|PCDH 3X|PCDH beta16|PCDH-beta-16|PCDH3X|PCDHB 16|PCDHB 8a|PCDHB8a|PCDHbeta 16|Protocadherin 3x|Protocadherin beta 16|Protocadherin beta 8a|Protocadherin beta-16|Protocadherin-3X|Protocadherin3x
- Cadherin ME1|KIAA1621|ME 1|ME1|PCDBG_HUMAN|PCDH 3X|PCDH beta16|PCDH-beta-16|PCDH3X|PCDHB 16|PCDHB 8a|PCDHB8a|PCDHbeta 16|Protocadherin 3x|Protocadherin beta 16|Protocadherin beta 8a|Protocadherin beta-16|Protocadherin-3X|Protocadherin3x
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant Human Protocadherin beta-16 protein (215-485AA)
- Recombinant Human Protocadherin beta-16 protein (215-485AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice