Anti-SLC39A10 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: SLC39A10
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77844-483
- 77844-482
- Antigen Symbol:
- SLC39A10
- SLC39A10
- Antigen Synonyms:
- 2900042E17Rik| 5430433I10| DKFZp781L10106| KIAA1265| LZT-Hs2| MGC126565| MGC138428| mKIAA1265| OTTMUSP00000023810| S39AA_HUMAN| slc39a10| Solute carrier family 39 (metal ion transporter), member 10| Solute carrier family 39 (zinc transporter), member 10| Solute carrier family 39 member 10| Zinc transporter ZIP10| ZIP-10| ZIP10| Zrt- and Irt-like protein 10
- 2900042E17Rik| 5430433I10| DKFZp781L10106| KIAA1265| LZT-Hs2| MGC126565| MGC138428| mKIAA1265| OTTMUSP00000023810| S39AA_HUMAN| slc39a10| Solute carrier family 39 (metal ion transporter), member 10| Solute carrier family 39 (zinc transporter), member 10| Solute carrier family 39 member 10| Zinc transporter ZIP10| ZIP-10| ZIP10| Zrt- and Irt-like protein 10
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9ULF5
- Q9ULF5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Zinc transporter ZIP10 protein (41-317AA)
- Recombinant Human Zinc transporter ZIP10 protein (41-317AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice