Anti-LRRC4C Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: LRRC4C
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77843-282
- 77843-283
- Antigen Symbol:
- LRRC4C
- LRRC4C
- Antigen Synonyms:
- 6430556C10Rik|KIAA1580|Leucine rich repeat containing 4C|Leucine-rich repeat-containing protein 4C|LRC4C_HUMAN|mKIAA1580|Netrin-G1 ligand|NGL 1|NGL-1|NGL1|RGD1311013
- 6430556C10Rik|KIAA1580|Leucine rich repeat containing 4C|Leucine-rich repeat-containing protein 4C|LRC4C_HUMAN|mKIAA1580|Netrin-G1 ligand|NGL 1|NGL-1|NGL1|RGD1311013
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9HCJ2
- Q9HCJ2
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Immunogen:
- Recombinant human Leucine-rich repeat-containing protein 4C protein (201-500AA)
- Recombinant human Leucine-rich repeat-containing protein 4C protein (201-500AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice