Anti-CACNB1 Rabbit Recombinant Polyclonal Antibody
77852-258
New Product
- Antigen Symbol:CACNB1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q02641
- Antigen Synonyms:CAB1|CACB1_HUMAN|CACNLB1|Calcium channel L type beta 1 polypeptide|Calcium channel voltage dependent beta 1 subunit|Calcium channel voltage-dependent subunit beta 1|CCHLB1|Dihydropyridine sensitive L type calcium channel beta 1 subunit|MGC41896|Voltage dependent L type calcium channel beta 1 subunit|Voltage-dependent L-type calcium channel subunit beta-1
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Voltage-dependent L-type calcium channel subunit beta-1 protein (429-598AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: CACNB1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human