Anti-PIP5K1A Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:200 to 1:1000, IHC: 1:25 to 1:100
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77848-835
- 77848-834
- Antigen Symbol:
- PIP5K1A
- PIP5K1A
- Antigen Synonyms:
- 68 kDa type I phosphatidylinositol-4-phosphate 5-kinase alpha|Phosphatidylinositol 4 phosphate 5 kinase type I alpha|Phosphatidylinositol-4-phosphate 5-kinase type I alpha|Phosphatidylinositol-4-phosphate 5-kinase type-1 alpha|PI4P5K Ia|PI51A_HUMAN|PIP5K A|PIP5K1-alpha|Pip5k1a|PIP5KIalpha|PtdIns(4)P-5-kinase 1 alpha
- 68 kDa type I phosphatidylinositol-4-phosphate 5-kinase alpha|Phosphatidylinositol 4 phosphate 5 kinase type I alpha|Phosphatidylinositol-4-phosphate 5-kinase type I alpha|Phosphatidylinositol-4-phosphate 5-kinase type-1 alpha|PI4P5K Ia|PI51A_HUMAN|PIP5K A|PIP5K1-alpha|Pip5k1a|PIP5KIalpha|PtdIns(4)P-5-kinase 1 alpha
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q99755
- Q99755
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human PIP5K1A
- Fusion protein of Human PIP5K1A
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice