Anti-VPS29 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: VPS29
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-762
- 77843-761
- Antigen Symbol:
- VPS29
- VPS29
- Antigen Synonyms:
- DC15|DC7|DKFZp564F0223|FLJ20492|hPEP11|PEP11 homolog|Retromer protein|Vacuolar protein sorting 29|Vacuolar protein sorting 29 (yeast homolog)|Vacuolar protein sorting 29 (yeast)|Vacuolar protein sorting 29 homolog (S. cerevisiae)|Vacuolar protein sorting 29 homolog|Vacuolar protein sorting associated protein 29|Vacuolar protein sorting-associated protein 29|Vacuolar sorting protein VPS29/PEP11|Vesicle protein sorting 29|VPS 29|VPS29 retromer complex component|VPS29_HUMAN|x 007 protein
- DC15|DC7|DKFZp564F0223|FLJ20492|hPEP11|PEP11 homolog|Retromer protein|Vacuolar protein sorting 29|Vacuolar protein sorting 29 (yeast homolog)|Vacuolar protein sorting 29 (yeast)|Vacuolar protein sorting 29 homolog (S. cerevisiae)|Vacuolar protein sorting 29 homolog|Vacuolar protein sorting associated protein 29|Vacuolar protein sorting-associated protein 29|Vacuolar sorting protein VPS29/PEP11|Vesicle protein sorting 29|VPS 29|VPS29 retromer complex component|VPS29_HUMAN|x 007 protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UBQ0
- Q9UBQ0
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Vacuolar protein sorting-associated protein 29 protein (68-145AA)
- Recombinant Human Vacuolar protein sorting-associated protein 29 protein (68-145AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice