Anti-ACTRT1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77850-699
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ACTRT1
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:Q8TDG2
- Antigen Synonyms:ACTRT1|ARPT1|HSD27Actin-related protein T1|ARP-T1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Actin-related protein T1 protein (201-376AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: ACTRT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human