Anti-ACP6 Rabbit Recombinant Polyclonal Antibody
Catalog # 77843-369
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ACP6
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q9NPH0
- Antigen Synonyms:Acid phosphatase 6|lysophosphatidic|Acid phosphatase like 1|Acid phosphatase like protein 1|Acid phosphatase-like protein 1|ACPL1|LPAP|Lysophosphatidic acid phosphatase 6|Lysophosphatidic acid phosphatase type 6|Lysophosphatidic acid phosphatase type 6 precursor|PACPL1|PPA6_HUMAN
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Lysophosphatidic acid phosphatase type 6 protein (35-324AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: ACP6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat