Anti-RPS6KA6 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:200 to 1:500
Type:
Antigen: RPS6KA6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-588
- 77844-587
- Antigen Symbol:
- RPS6KA6
- RPS6KA6
- Antigen Synonyms:
- 90 kDa ribosomal protein S6 kinase 6|KS6A6_HUMAN|p90 RSK 6|p90-RSK 6|p90RSK6|pp90RSK4|Ribosomal protein S6 kinase 90kDa polypeptide 6|Ribosomal protein S6 kinase alpha 6|Ribosomal protein S6 kinase alpha-6|Ribosomal S6 kinase 4|RSK 4|RSK-4|S6K-alpha-6
- 90 kDa ribosomal protein S6 kinase 6|KS6A6_HUMAN|p90 RSK 6|p90-RSK 6|p90RSK6|pp90RSK4|Ribosomal protein S6 kinase 90kDa polypeptide 6|Ribosomal protein S6 kinase alpha 6|Ribosomal protein S6 kinase alpha-6|Ribosomal S6 kinase 4|RSK 4|RSK-4|S6K-alpha-6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UK32
- Q9UK32
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant human Ribosomal protein S6 kinase alpha-6 protein (340-467AA)
- Recombinant human Ribosomal protein S6 kinase alpha-6 protein (340-467AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice