Anti-CYFIP2 Rabbit Polyclonal Antibody
Catalog # 77848-558
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CYFIP2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q96F07
- Antigen Synonyms:KIAA1168|PIR121Cytoplasmic FMR1-interacting protein 2|p53-inducible protein 121
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from CYFIP2 at AA range: 1171-1220
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 2000, ELISA:1:10000 to 20000
Type:
Antigen: CYFIP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat